EMAIL THIS PAGE TO A FRIEND
S7004 Sigma-Aldrich

Sphingomyelin

from bovine brain, ≥97.0%, powder

Synonym: N-Acyl-4-sphingenyl-1-O-phosphorylcholine, N-Acyl-D-sphingosine-1-phosphocholine

Purchase

Properties

biological source   bovine brain
assay   ≥97.0%
form   powder
color   white
storage temp.   −20°C

Description

Application

Sphingomyelin was used as exogenous treatment to study the effect on ATP depletion and Fe-mediated cell injury in human proximal tubular cell line, HK-2 cells.3 It was used to study the effect on Akt phosphorylation in obese mice.4

Biochem/physiol Actions

Sphingomyelin (SM) is a sphingolipid found in the myelin sheath and cell membranes of animals. The regulation of SM content by sphingomyelinase and SM synthase 2 affect the membrane properties and signaling.1 Excess oxysterols result in increased amount of SM and calcium ions that is the main reason for artery blockage and atherosclerosis.2

Other Notes

Contains primarily stearic, nevonic, and lignoceric acids.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles

Articles

LC/MS Analysis of Phospholipids in Milk on Ascentis® Express HILIC after SPE using Supelclean™ Si, Shotgun HPLC Analysis

From our library of Articles, Sigma-Aldrich presents LC/MS Analysis of Phospholipids in Milk on Ascentis® Express HILIC after SPE using Supelclean™ Si, Shotgun HPLC Analysis
Keywords: Chromatography, High performance liquid chromatography, Solid phase extractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?