EMAIL THIS PAGE TO A FRIEND
S8143 Sigma-Aldrich

D-Sorbitol

suitable for plant cell culture

Synonym: D-Glucitol

  • CAS Number 50-70-4

  • Empirical Formula (Hill Notation) C6H14O6

  • Molecular Weight 182.17

  •  Beilstein/REAXYS Number 1721899

  •  EC Number 200-061-5

  •  MDL number MFCD00004708

  •  PubChem Substance ID 24899777

Purchase

Properties

Related Categories Carbohydrate Chemistry, Carbohydrate Derivatives, Carbohydrates, Cell Biology, Chemical Biology,
vapor density   <1 (vs air)
vapor pressure   <0.1 mmHg ( 25 °C)
InChI Key   FBPFZTCFMRRESA-JGWLITMVSA-N
assay   ≥98% (GC)
form   powder
application(s)   cell culture | plant: suitable
mp   98-100 °C (lit.)

Description

Application

May be used for washing spheroplasts and in isoelectric focusing to minimize endoosmotic flow in agarose gels. May be used to induce osmotic stress.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg
RTECS 
LZ4290000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number

Single- & multi-channel pipettes from BRAND
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?