• USA Home
  • S8628 - Sodium cyanoborohydride

EMAIL THIS PAGE TO A FRIEND
S8628 Sigma-Aldrich

Sodium cyanoborohydride

≥90% (titration)

Synonym: Sodium cyanotrihydridoborate

  • CAS Number 25895-60-7

  • Linear Formula NaBH3CN

  • Molecular Weight 62.84

  •  Beilstein/REAXYS Number 4152656

  •  MDL number MFCD00003516

Purchase

Properties

assay   ≥90% (titration)
mp   >242 °C (dec.) (lit.)
solubility   H2O: soluble, clear to slightly hazy
SMILES string   [Na+].[BH3-]C#N
InChI   1S/CH3BN.Na/c2-1-3;/h2H3;/q-1;+1
InChI key   CVDUGUOQTVTBJH-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Signal word 
Danger
Supplemental Hazard Statements 
EUH032
RIDADR 
UN 3179 6.1(4.1) / PGII
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

156159 Sodium cyanoborohydride, reagent grade, 95%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?