EMAIL THIS PAGE TO A FRIEND
S9148 Sigma

Seawater

Untreated, suitable for plant cell culture

Purchase

Properties

Related Categories Algae Culture, BioFuels, Cell Biology, Molecular Biology, Phycology and Ecotoxicity Products,
Quality Level   GMP
sterility   sterile-filtered
composition   Salinity, ≥32 ppt
suitability   suitable for plant cell culture
Featured Industry   Agriculture

Description

Preparation Note

Natural, collected from Gulf Stream in the Gulf of Mexico.

Application

Seawater S9148 is collected from the Gulf Steam in the Gulf of Mexico and membrane-filtered for use in the reconstitution of marine cell culture media such as Guillard’s (F/2) Marine media. Guillard’s (F/2) Marine Enrichment medium is a widely used enriched seawater medium for growing costal marine algae (diatoms).

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis


Request a Serum Sample

The Emprove® Program
Protocols & Articles

Related Content

Synthetic Biology Resources

Sigma® Life Science is working to use our stregnths to build the field of Synthetic Biology. Below is a collection of links to our favorite resources. Sigma also offers thousands of products for your...
Keywords: Cell culture, Cloning, Gene expression, Genetic, Genetics, Metabolic Pathways, Metabolites, Molecular biology, Molecular biology techniques, Photosynthesis, Poisons, Polymerase chain reaction - quantitative, Transcription, Transfection

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?