• USA Home
  • SAB1400656 - Anti-RPTOR antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB1400656 Sigma-Aldrich

Anti-RPTOR antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-KIAA1303

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RAE-RAX
species reactivity   human
application(s)   indirect immunofluorescence: suitable
  western blot: 1 μg/mL
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... RPTOR(57521)
biological source   mouse
antibody product type   primary antibodies
conjugate   unconjugated
UniProt accession no.   Q8N122

Description

Immunogen

RPTOR (AAH64515.1, 1 a.a. ~ 379 a.a) full-length human protein.

Sequence
MESEMLQSPLLGLGEEDEADLTDWNLPLAFMKKRHCEKIEGSKSLAQSWRMKDRMKTVSVALVLCLNVGVDPPDVVKTTPCARLECWIDPLSMGPQKALETIGANLQKQYENWQPRARYKQSLDPTVDEVKKLCTSLRRNAKEERVLFHYNGHGVPRPTVNGEVWVFNKNYTQYIPLSIYDLQTWMGSPSIFVYDCSNAGLIVKSFKQFALQREQELEVAAINPNHPLAQMPLPPSMKNCIQLAACEATELLPMIPDLPADLFTSCLTTPIKIALRWFCMQKCVSLVPGVTLDLIEKIPGRLNDRRTPLGELNWIFTAITDTIAWNVLPRDLFQKLFRQDLLVASLFRNFLLAERIMRSYNCTPVSSPRLPPTYMHAMW

General description

This gene encodes a component of a signaling pathway that regulates cell growth in response to nutrient and insulin levels. The encoded protein forms a stoichiometric complex with the mTOR kinase, and also associates with eukaryotic initiation factor 4E-binding protein-1 and ribosomal protein S6 kinase. The protein positively regulates the downstream effector ribosomal protein S6 kinase, and negatively regulates the mTOR kinase. Multiple transcript variants encoding different isoforms have been found for this gene. (provided by RefSeq)

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?