• USA Home
  • SAB1407967 - Anti-MCTP1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB1407967 Sigma-Aldrich

Anti-MCTP1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym: FLJ22344

  •  NACRES NA.41

Purchase

Properties

conjugate   unconjugated
clone   polyclonal
biological source   mouse
application(s)   western blot: 1 μg/mL
species reactivity   human
mol wt   antigen ~68.5 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   BC030005.2
UniProt accession no.   Q6DN14
Gene Information   human ... MCTP1(79772)

Description

Immunogen

MCTP1 (AAH30005.1, 1 a.a. ~ 600 a.a) full-length human protein.

Sequence
MLDSCKLKSACNLPFICNKKIINTAGTSNAEVPLADPGMYQLDITLRRGQSLAARDRGGTSDPYVKFKIGGKEVFRSKIIHKNLNPVWEEKACILVDHLREPLYIKVFDYDFGLQDDFMGSAFLDLTQLELNRPTDVTLTLKDPHYPDHDLGIILLSVILTPKEGESRDVTMLMRKSWKRSSKELSENEVVGSYFSVKSLFWRTCGRPALPVLGFCRAELQNPYCKNVQFQTQSLRLSDLHRKSHLWRGIVSITLIEGRDLKAMDSNGLSDPYVKFRLGHQKYKSKIMPKTLNPQWREQFDFHLYEERGGVIDITAWDKDAGKRDDFIGRCQVDLSALSREQTHKLELQLEEGEGHLVLLVTLTASATVSISDLSVNSLEDQKEREEILKKYSPLRIFHNLKDVGFLQVKVIRAEGLMAADVTGKSDPFCVVELNNDRLLTHTVYKNLNPEWNKVFTFNIKDIHSVLEVTVYDEDRDRSADFLGKVAIPLLSIQNGEQKAYVLKNKQLTGPTKGVIYLEIDVIFNAVKASLRTLIPKEQKYIEEENRLSKQLLLRNFIRMKRCVMVLVNAAYYVNSCFDWDSPPRSLAAFVALYGTFINV

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?