• USA Home
  • SAB1408188 - Anti-PCDH21 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB1408188 Sigma-Aldrich

Anti-PCDH21 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym: DKFZp434A132, KIAA1775, PRCAD

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PB-PC, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   mouse
application(s)   western blot: 1 μg/mL
species reactivity   human
mol wt   antigen mol wt ~81.5 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
antibody product type   primary antibodies
NCBI accession no.   BC038799.1
Gene Information   human ... PCDH21(92211)

Description

General description

This gene is a member of the cadherin superfamily of calcium-dependent cell-cell adhesion molecules. The encoded protein has a signal peptide, six cadherin repeat domains and a unique cytoplasmic region. This non-classical cadherin appears to be exclusively expressed in the mitral and tufted cells in the main and accessory olfactory bulbs of the brain, suggesting a possible role in the formation and maintenance of neuronal networks. (provided by RefSeq)

Immunogen

PCDH21 (AAH38799.1, 1 a.a. ~ 745 a.a) full-length human protein.

Sequence
MRRCRWAALALGLLRLCLAQANFAPHFFDNGVGSTNGNMALFSLPEDTPVGSHVYTLNGTDPEGDPISYHISFDPSTRSVFSVDPTFGNITLVEELDREREDEIEAIISISDGLNLVAEKVVILVTDANDEAPRFIQEPYVALVPEDIPAGSIIFKVHAVDRDTGSGGSVTYFLQNLHSPFAVDRHSGVLRLQAGATLDYERSRTHYITVVAKDGGGRLHGADVVFSATTTVTVNVEDVQDMAPVFVGTPYYGYVYEDTLPGSEVLKVVAMDGDRGKPNRILYSLVNGNDGAFEINETSGAISITQSPAQLQREVYELHVQVTEMSPAGSPAAQATVPVTIRIVDLNNHPPTFYGESGPQNRFELSMNEHPPQGEILRGLKITVNDSDQGANAKFNLQLVGPRGIFRVVPQTVLNEAQVTIIVENSAAIDFEKSKVLTFKLLAVEVNTPEKFSSTADVVIQLLDTNDNVPKFDSLYYVARIPENAPGGSSVVAVTAVDPDTGPWGEVKYSTYGTGADLFLIHPSTGLIYTQPWASLDAEATARYNFYVKAEDMEGKYSVAEVFITLLDVNDHPPQFGKSVQKKTMVLGTPVKIEAIDEDAEEPNNLVDYSITHAEPANVFDINSHTGEIWLKNSIRSLDALHNITPGRDCLWSLEVQAKDRGSPSFSTTALLKIDITDAEVRRLRYMKNSNFPGTTKSVRKPKFKPKKPHSSQGLFLHPHCEIALFNLSNVNLYSRVFQGAAQAS

Physical form

Solution in phosphate buffered saline, pH 7.4

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?