• USA Home
  • SAB1408383 - Anti-PLB1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB1408383 Sigma-Aldrich

Anti-PLB1 antibody produced in mouse

purified immunoglobulin, buffered aqueous solution

Synonym: PLB

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PJ-PN, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   mouse
application(s)   western blot: 1 μg/mL
species reactivity   human
mol wt   antigen ~54.3 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   BC065041
UniProt accession no.   Q6P1J6
Gene Information   human ... PLB1(151056)

Description

Immunogen

PLB1 (AAH65041, 1 a.a. ~ 488 a.a) full-length human protein.

Sequence
MEPAGEKDEPLSVKHGRPMKCPSQESPYLFSYRNSNYLTRLQKPQDKLEVREGAEIRCPDKDPSDTVPTSVHRLKPADINVIGALGDSLTAGNGAGSTPGNVLDVLTQYRGLSWSVGGDENIGTVTTLANILREFNPSLKGFSVGTGKETSPNAFLNQAVAGGRAEDLPVQARRLVDLMKNDTRIHFQEDWKIITLFIGGNDLCDFCNDLVHYSPQNFTDNIGKALDILHAEVPRAFVNLVTVLEIVNLRELYQEKKVYCPRMILRSLCPCVLKFDDNSTELATLIEFNKKFQEKTHQLIESGRYDTREDFTVVVQPFFENVDMPKTSEGLPDNSFFAPDCFHFSSKSHSRAASALWNNMLEPVGQKTTRHKFENKINITCPNQVQPFLRTYKNSMQGHGTWLPCRDRAPSALHPTSVHALRPADIQVVAALGDSLTAGNGIGSKPDDLPDVTTQYRGLSYRESKPGFLSDSWVSKSNRKCTRKAPNP

Physical form

Solution in phosphate buffered saline, pH 7.4

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?