• USA Home
  • SAB1410662 - Anti-HSD17B2 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1410662 Sigma-Aldrich

Anti-HSD17B2 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym: EDH17B2, HSD17, SDR9C2

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, HP-HT, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 1 μg/mL
species reactivity   human, mouse
mol wt   antigen 42.8 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   NM_002153.1
UniProt accession no.   P37059
Gene Information   human ... HSD17B2(3294)

Description

General description

HSD17B2 (Hydroxysteroid 17-β dehydrogenase 2) is a member of the short chain alcohol dehydrogenase superfamily expressed in non-steroidogenic human embryonic kidney cells.

Immunogen

HSD17B2 (NP_002144.1, 1 a.a. ~ 387 a.a) full-length human protein.

Sequence
MSTFFSDTAWICLAVPTVLCGTVFCKYKKSSGQLWSWMVCLAGLCAVCLLILSPFWGLILFSVSCFLMYTYLSGQELLPVDQKAVLVTGGDCGLGHALCKYLDELGFTVFAGVLNENGPGAEELRRTCSPRLSVLQMDITKPVQIKDAYSKVAAMLQDRGLWAVINNAGVLGFPTDGELLLMTDYKQCMAVNFFGTVEVTKTFLPLLRKSKGRLVNVSSMGGGAPMERLASYGSSKAAVTMFSSVMRLELSKWGIKVASIQPGGFLTNIAGTSDKWEKLEKDILDHLPAEVQEDYGQDYILAQRNFLLLINSLASKDFSPVLRDIQHAILAKSPFAYYTPGKGAYLWICLAHYLPIGIYDYFAKRHFGQDKPMPRALRMPNYKKKAT

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

HSD17B2 (Hydroxysteroid 17-β dehydrogenase 2) is highly involved in the synthesis and metabolism of steroid hormones. It maintains the intracellular concentration of biologically active steroid hormones in a wide range of male and female human tissues. It catalyzes the interconversion of testosterone and androstenedione as well as estradiol and estrone through 17-β HSD activities. HSD17B2 also exhibits of 20α-HSD activity toward 20α-dihydroprogesterone.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?