• USA Home
  • SAB1411734 - Monoclonal Anti-HA antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB1411734 Sigma-Aldrich

Monoclonal Anti-HA antibody produced in mouse

clone 4D4, purified immunoglobulin, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, H1-HE, Primary Antibodies
conjugate   unconjugated
clone   4D4, monoclonal
biological source   mouse
application(s)   indirect ELISA: suitable
species reactivity   human
mol wt   antigen 75 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1κ
antibody product type   primary antibodies
NCBI accession no.   AY651336.1
Gene Information   human ... HA(3654620)

Description

General description

Influenza virus hemagglutinin (HA) precursor is encoded by the gene mapped on segment 4. The encoded protein is a 76KDa surface glycoprotein and it exists as trimers on the envelope.

Immunogen

Recombinant Fc fusion protein corresponding to amino acids 17-531 of virus HA (Influenza A virus, A/Viet Nam/3062/2004 (H5N1))

Sequence
DQICIGYHANNSTEQVDTIMEKNVTVTHAQDILEKTHNGKLCDLDGVKPLILRDCSVAGWLLGNPMCDEFINVPEWSYIVEKANPVNDLCYPGDFNDYEELKHLLSRINHFEKIQIIPKSSWSSHEASLGVSSACPYQGKSSFFRNVVWLIKKNSTYPTIKRSYNNTNQEDLLVLWGIHHPNDAAEQTKLYQNPTTYISVGTSTLNQRLVPRIATRSKVNGQSGRMEFFWTILKPNDAINFESNGNFIAPEYAYKIVKKGDSTIMKSELEYGNCNTKCQTPMGAINSSMPFHNIHPLTIGECPKYVKSNRLVLATGLRNSPQRERRRKKRGLFGAIAGFIEGGWQGMVDGWYGYHHSNEQGSGYAADKESTQKAIDGVTNKVNSIIDKMNTQFEAVGREFNNLERRIENLNKKMEDGFLDVWTYNAELLVLMENERTLDFHDSNVKNLYDKVRLQLRDNAKELGNGCFEFYHKCDNECMESVRNGTYDYPQYSEEARLKREEISGVKLESIGIYQ

Physical form

Solution in phosphate buffered saline, pH 7.4

Biochem/physiol Actions

Influenza virus hemagglutinin (HA) acts as a key immunogen in avian influenza virus (AIV) vaccines. Activated HA facilitates virus attachment to oligosaccharide receptors on host cell surface and is involved in penetration through fusion following conformational modifications in the endolysosomal compartment. 226 and 228 residues in hemagglutinin (HA) of the H3 subtype plays a key role in host range restriction and receptor specificity. Influenza virus hemagglutinin (HA) is responsible for 1918 influenza pandemic.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?