• USA Home
  • SAB1413081 - Monoclonal Anti-RELA (phospho S536) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1413081 Sigma-Aldrich

Monoclonal Anti-RELA (phospho S536) antibody produced in rabbit

New

clone B7

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RB-RG
clone   B7, monoclonal
biological source   rabbit
application(s)   flow cytometry: suitable
  flow cytometry: suitable
  western blot: suitable
species reactivity   human
form   buffered aqueous glycerol solution
storage temp.   −20°C
antibody product type   primary antibodies
Gene Information   human ... RELA(5970)

Description

Immunogen

A synthetic phosphopeptide corresponding to residues surrounding S536 of human RELA.

Physical form

In PBS, pH 7.4 (50% glycerol, 0.1% BSA and 0.02% sodium azide).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?