• USA Home
  • SAB2101402 - Anti-LST-3TM12 (ab2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2101402 Sigma-Aldrich

Anti-LST-3TM12 (ab2) antibody produced in rabbit

affinity isolated antibody

Synonym: Anti-LST3, Anti-Organic anion transporter LST-3b

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Drug Transport Proteins, Biochemicals and Reagents, Drug Transport Proteins & Reagents,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: suitable
species reactivity   human
mol wt   71 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   0.5 mg - 1 mg/mL
UniProt accession no.   Q71QF0
Gene Information   human ... LST-3TM12(338821)

Description

Immunogen

Synthetic peptide directed towards the middle region of human LST-3TM12

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Sequence

Synthetic peptide located within the following region: LKTNDKRNQIANLTNRRKYITKNVTGFFQSLKSILTNPLYVIFVIFTLLH

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Drug Transport

Protein-based drug transporters are found in most tissues including liver, kidney, intestine, and brain. Because of their complexity and genetic heterogeneity, these proteins are often produced as re...
Robert Gates
Biofiles Volume 6 Article 1
Keywords: Absorption, Anti-inflammatory agents, Antibiotics, Antimicrobials, Apoptosis, Cancer, Catalog, Cytotoxins, Detoxification, Enzyme-linked immunosorbent assay, Gene expression, Genetic, High performance liquid chromatography, Immunohistochemistry, Mass spectrometry, Metabolites, Methods, Phosphorylations, Separation, The Reporter, Transfection, Type, Western blot

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?