• USA Home
  • SAB2102728 - Anti-XRCC4 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2102728 Sigma

Anti-XRCC4 antibody produced in rabbit

affinity isolated antibody

Synonym: Anti-X-ray repair complementing defective repair in Chinese hamster cells 4

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Common Transcription Factors, Antibodies to Transcription Factors, Epigenetics,
species reactivity   human, horse, bovine, guinea pig
application(s)   western blot: suitable
clone   polyclonal
concentration   0.5 mg - 1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   mol wt 38 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... XRCC4(7518)
biological source   rabbit
antibody product type   primary antibodies
conjugate   unconjugated
UniProt accession no.   Q13426

Description

Immunogen

Synthetic peptide directed towards the middle region of human XRCC4

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

XRCC4 functions together with DNA ligase IV and the DNA-dependent protein kinase in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. The non-homologous end-joining pathway is required both for normal development and for suppression of tumors. This gene functionally complements XR-1 Chinese hamster ovary cell mutant, which is impaired in DNA double-strand breaks produced by ionizing radiation and restriction enzymes.The protein encoded by this gene functions together with DNA ligase IV and the DNA-dependent protein kinase in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. The non-homologous end-joining pathway is required both for normal development and for suppression of tumors. This gene functionally complements XR-1 Chinese hamster ovary cell mutant, which is impaired in DNA double-strand breaks produced by ionizing radiation and restriction enzymes. This gene contains 8 exons, and alternative transcription initiation and alternative splicing generates several transcript variants.

Sequence

Synthetic peptide located within the following region: LQKENERLLRDWNDVQGRFEKCVSAKEALETDLYKRFILVLNEKKTKIRS

Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Amplification, Buffers, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?