• USA Home
  • SAB2500893 - Anti-ROBO3/RIG1 (Internal) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
SAB2500893 Sigma-Aldrich

Anti-ROBO3/RIG1 (Internal) antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-FLJ21044, Anti-RIG1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RO-RP
conjugate   unconjugated
clone   polyclonal
biological source   goat
application(s)   indirect ELISA: suitable
  western blot: suitable
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   Q96MS0
Gene Information   human ... ROBO3(64221)

Description

Immunogen

Peptide with sequence C-RRKQRKELSHYT from the internal region of the protein sequence according to NP_071765.2.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?