• USA Home
  • SAB2700878 - Anti-IL17RC antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2700878 Sigma-Aldrich

Anti-IL17RC antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: FLJ95963, FLJ96005, IL17-RL, IL17RC

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, IH-IM, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: 1:500-1:3000
species reactivity   human
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   NP_703191
UniProt accession no.   Q8NAC3
Gene Information   human ... IL17RC(84818)

Description

General description

IL17RC (Interleukin 17 receptor C) is a novel single-pass transmembrane protein belonging to the interleukin-17 cytokine family. It is expressed in T cells and B cells with an extracellular N terminal domain. It is widely distributed in prostate, cartilage, kidney, liver, heart, and muscle, whereas at a lower level in thymus and leukocytes. It is mapped on human chromosome 3p25.3-3p24.1. Its expression has also been found in various cancerous cells such as prostate cancer.

Immunogen

Recombinant protein fragment contain a sequence corresponding to a region within amino acids 62 and 471 of Human Interleukin-17 receptor C according to NP_703191

Application

Anti-IL17RC antibody produced in rabbit is suitable for western blot anaysis.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Biochem/physiol Actions

IL17RC (Interleukin 17 receptor C) performs in the cytokine signaling pathway as a decoy receptor. It is essentially required for the signaling of IL-17A, IL-17F, and IL-17A/F both in vitro and in vivo. In nonhemopoietic tissues, it couples with IL-17RA and forms a complex, which drives the functions of IL-17A and IL-17F homodimers as well as IL-17A/F heterodimers. Inherited deficiency of IL17RC causes an autosomal recessive disorder, chronic mucocutaneous candidiasis (CMC), with recurrent or persistent infections of the skin, nail, oral, and genital mucosae.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?