• USA Home
  • SAB2701036 - Anti-LHX8 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2701036 Sigma-Aldrich

Anti-LHX8 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: LHX8, Lhx7

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, L-LL, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunoprecipitation (IP): suitable
  western blot: 1000-10000
species reactivity   human
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   NP_001001933
UniProt accession no.   Q68G74
Gene Information   human ... LHX8(431707)

Description

Immunogen

Recombinant fragment corresponding to a region within amino acids 1 and 356 of LIM homeobox protein 8 according to NP_001001933

Application

Suggested starting dilutions are as follows: IP: 1:100-1:500, WB: 1:1000-1:10000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Physical form

0.1M Tris, 0.1M Glycine, 10% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Biochem/physiol Actions

Members of the LIM homeobox gene family, such as LHX8, encode transcription regulators that share common structural features. They all contain 2 tandemly repeated cysteine-rich double-zinc finger motifs, called LIM domains, in addition to a homeodomain. The homeodomain is a DNA-binding domain, and the LIM domains are essential for regulating the activity of these molecules by interacting with other proteins. Members of the LIM homeobox gene family are required for the patterning or the specification and differentiation of different cell types during embryonic development (Zhao et al., 1999 [PubMed 10611327]).[supplied by OMIM]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?