• USA Home
  • SAB2701473 - Anti-PSG3 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2701473 Sigma-Aldrich

Anti-PSG3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: PSG3, pregnancy specific beta-1-glycoprotein 3

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PS-PS, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry: suitable
  western blot: 500-3000
species reactivity   human
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
NCBI accession no.   AAH05924
UniProt accession no.   Q16557
Gene Information   human ... PSG3(5671)

Description

Immunogen

Recombinant fragment corresponding to a region within amino acids 11 and 347 of PSG3 according to AAH05924

Application

Suggested starting dilutions are as follows: IHC-P: 1:100-1:1000, WB: 1:500-1:3000. Not yet tested in other applications. Optimal working dilutions should be determined experimentally by the end user.

Physical form

0.1M Tris, 0.1M Glycine, 20% Glycerol (pH7). 0.01% Thimerosal was added as a preservative.

Biochem/physiol Actions

The human pregnancy-specific glycoproteins (PSGs) are a family of proteins that are synthesized in large amounts by placental trophoblasts and released into the maternal circulation during pregnancy. Molecular cloning and analysis of several PSG genes has indicated that the PSGs form a subgroup of the carcinoembryonic antigen (CEA) gene family, which belongs to the immunoglobulin superfamily of genes. Members of the CEA family consist of a single N domain, with structural similarity to the immunoglobulin variable domains, followed by a variable number of immunoglobulin constant-like A and/or B domains. Most PSGs have an arg-gly-asp (RGD) motif, which has been shown to function as an adhesion recognition signal for several integrins, in the N-terminal domain (summary by Teglund et al., 1994 [PubMed 7851896]). For additional general information about the PSG gene family, see PSG1 (MIM 176390).[supplied by OMIM]

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?