• USA Home
  • SAB2701860 - Anti-actn1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2701860 Sigma-Aldrich

Anti-actn1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: actinin, alpha 1, actn1

  •  NACRES NA.41

Purchase

Properties

Related Categories AC-AC, Alphabetical Index, Antibodies, Cell Biology, Markers and Controls,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   western blot: suitable
species reactivity   zebrafish
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
Quality Level   100
antibody product type   primary antibodies
NCBI accession no.   NP_001161758
Gene Information   zebrafish ... actn1(560400)

Description

Immunogen

Recombinant protein containing a sequence corresponding to a region within amino acids 61 and 227 of actn1 according to NP_001161758

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?