• USA Home
  • SAB2900055 - Anti-EPHA4 (AB2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2900055 Sigma-Aldrich

Anti-EPHA4 (AB2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: CEK8, HEK8, eph receptor a4, ephrin type-a receptor 4, sek

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, EO-ER, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
species reactivity   human
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
UniProt accession no.   P54764
Gene Information   human ... EPHA4(2043)

Description

Immunogen

Synthetic 16 amino acid peptide from internal region of human EPHA4. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Marmoset (100%); Dog, Panda, Horse (94%); Bovine, Bat, Hamster, Elephant, Pig, Opossum, Turkey, Chicken (88%); Mouse, Rat, Rabbit, Platypus, Lizard, Xenopus (81%).

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?