• USA Home
  • SAB3300090 - Monoclonal Anti-GATA4 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB3300090 Sigma-Aldrich

Monoclonal Anti-GATA4 antibody produced in mouse

clone 9F9, purified immunoglobulin, buffered aqueous glycerol solution

Synonym: Anti-MGC126629

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Proteins, Bioactives, Markers, and Receptors for Stem Cell Biology, Antibodies for Epigenetics and Nuclear Signaling, Antibodies for Gene Regulation,
conjugate   unconjugated
clone   9F9, monoclonal
biological source   mouse
application(s)   immunohistochemistry: suitable
  indirect immunofluorescence: suitable
  western blot: suitable
species reactivity   human
mol wt   antigen 44.4 kDa
form   buffered aqueous glycerol solution
shipped in   dry ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG3
antibody product type   primary antibodies
NCBI accession no.   NM_002052
UniProt accession no.   P43694
Gene Information   human ... GATA4(2626)

Description

Immunogen

Recombinant fragment expressed in E. coli corresponding to amino acids 296-442of human GATA4

Target description

GATA4 encodes a member of the GATA family of zinc-finger transcription factors. Members of this family recognize the GATA motif which is present in the promoters of many genes. This protein is thought to regulate genes involved in embryogenesis and in myocardial differentiation and function. Mutations in this gene have been associated with cardiac septal defects.

Physical form

Solution in phosphate buffered saline containing 1% bovine serum albumin, 50% glycerol, and 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?