• USA Home
  • SAB3501067 - Anti-ULK3 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3501067 Sigma-Aldrich

Anti-ULK3 antibody produced in rabbit

affinity isolated antibody

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides, Primary Antibodies, U-Z,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   ELISA: suitable
  immunoblotting: suitable
species reactivity   human
mol wt   predicted mol wt 52 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
Quality Level   100
antibody product type   primary antibodies
concentration   1 mg/mL
NCBI accession no.   NP_001092906
UniProt accession no.   Q6PHR2
Gene Information   human ... ULK3(25989)

Description

Immunogen

Antibody was raised against a 19 amino acid peptide near the center of human ULK3.

Physical form

Supplied at 1 mg/mL in PBS with 0.02% sodim azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3501067.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?