• USA Home
  • SAB3700276 - Anti-Goat IgG (H+L), highly cross adsorbed-Biotin antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3700276 Sigma-Aldrich

Anti-Goat IgG (H+L), highly cross adsorbed-Biotin antibody produced in rabbit

affinity isolated antibody, lyophilized powder

  •  NACRES NA.46

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Biotin Antibodies, GM-GO, Goat Secondary Antibodies and Conjugates,
conjugate   biotin conjugate
clone   polyclonal
biological source   rabbit
application(s)   immunohistochemistry: suitable
  indirect ELISA: suitable
  western blot: suitable
species reactivity   goat
form   lyophilized powder
shipped in   wet ice
storage temp.   2-8°C
antibody form   affinity isolated antibody
Quality Level   100
antibody product type   secondary antibodies

Description

General description

An immunoglobulin has two heavy chain and two light chain connected by disulfide bond. It mainly helps in immune defense. It is a glycoprotein. IgG is a major class of immunoglobulin. Immunoglobulin G (IgG) participates in hypersensitivity type II and type III. Goat IgG has two subclasses- IgG1 and IgG2.

Specificity

This product was prepared from monospecific antiserum by immunoaffinity chromatography using Goat IgG coupled to agarose beads followed by solid phase adsorption(s) to remove any unwanted reactivities. Assay by immunoelectrophoresis resulted in a single precipitin arc against Anti-Biotin, Anti-Rabbit Serum, Goat IgG and Goat Serum. No reaction was observed against Human Serum Proteins.

Immunogen

Goat IgG whole molecule

Physical form

Supplied in 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free

Reconstitution

Reconstitute with 1.0 mL deionized water (or equivalent).

Physical properties

Antibody format: IgG

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
EUH032
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

How to Choose a Secondary Antibody

The following information is provided to help you decide which secondary antibody may be best for your particular application.
Keywords: Amplification, Enzyme-linked immunosorbent assay, Flow cytometry, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western blot

Secondary Antibodies, Conjugates and Kits

Secondary antibodies are polyclonal or monoclonal antibodies that bind to primary antibodies or antibody fragments, such as the Fc or Fab regions. They are typically labeled with probes that make the...
Keywords: Absorption, Adsorption, Amplification, Bacterial conjugations, Cancer, Digestions, Electrophoresis, Enzyme-linked immunosorbent assay, Flow cytometry, Gel electrophoresis, Gene expression, Hormones, Immobilization, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunology, Immunoprecipitation, Infrared spectroscopy, Microbiology, Microscopy, Purification, Vitamins, Western blot

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?