• USA Home
  • SAB4100001 - Monoclonal Anti-AKT3 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4100001 Sigma-Aldrich

Monoclonal Anti-AKT3 antibody produced in mouse

clone AKT3-A1, culture supernatant

Synonym: Anti-PKB-γ, Anti-PRKBG, Anti-RAC-γ, Anti-RAC-PK-γ, Anti-STK-2, Anti-v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)

Purchase

Properties

conjugate   unconjugated
clone   AKT3-A1, monoclonal
biological source   mouse
application(s)   western blot: suitable
species reactivity   human
mol wt   antigen predicted mol wt 55.6 kDa
form   aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   culture supernatant
isotype   IgG3
antibody product type   primary antibodies
NCBI accession no.   NM_005465
UniProt accession no.   Q9Y243
Gene Information   human ... AKT3(10000)

Description

General description

AKT serine/threonine kinase 3 (AKT3) is part of the protein kinase B family. It has a kinase domain, a pleckstrin homology domain and a regulatory domain. The protein consists of 479 amino acids and the gene encoding it is localized on human chromosome 1q44.

Immunogen

AKT3 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

AKT serine/threonine kinase 3 (AKT3) has a role in DNA repair and brain development. In mouse model, it modulates the expression of estrogen receptor α, Erb-B2 receptor tyrosine kinase 2 and Erb-B2 receptor tyrosine kinase 3 in breast cancer cells. AKT3 has been shown to stimulate the growth of triple-negative breast cancer.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?