• USA Home
  • SAB4100133 - Monoclonal Anti-LDHA antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4100133 Sigma-Aldrich

Monoclonal Anti-LDHA antibody produced in mouse

clone LDHA-C47, culture supernatant

Synonym: Anti-Proliferation-inducing gene 19, Anti-GSD11, Anti-L-lactate dehydrogenase A, Anti-LDH1, Anti-LDHM, Anti-Lactate dehydrogenase A, Anti-PIG19

Purchase

Properties

Related Categories Aerobic Glycolysis (the Warburg Effect), Alphabetical Index, Antibodies, Antibodies Against Proteins Involved in Glucose Metabolism, Cancer Metabolism,
conjugate   unconjugated
clone   LDHA-C47, monoclonal
biological source   mouse
application(s)   western blot: suitable
species reactivity   human
mol wt   antigen predicted mol wt 36.5 kDa
form   aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   culture supernatant
isotype   IgG1
antibody product type   primary antibodies
UniProt accession no.   P00338
Gene Information   human ... LDHA(3939)

Description

Immunogen

LDHA purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary and Secondary Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay needs. Use the Antibody...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?