• USA Home
  • SAB4100140 - Monoclonal Anti-AK5 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4100140 Sigma-Aldrich

Monoclonal Anti-AK5 antibody produced in mouse

clone AK5-B105, culture supernatant

Synonym: Anti-OTTHUMP00000011354, Anti-OTTHUMP00000011355, Anti-AK6, Anti-ATP-AMP transphosphorylase, Anti-Adenylate kinase 5, Anti-Adenylate kinase 6

  •  NACRES NA.41

Purchase

Properties

Related Categories AE-AK, Alphabetical Index, Antibodies, Primary Antibodies
conjugate   unconjugated
clone   AK5-B105, monoclonal
biological source   mouse
application(s)   western blot: suitable
species reactivity   human
mol wt   antigen predicted mol wt 63.2 kDa
form   aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   culture supernatant
isotype   IgG2b
Quality Level   200
antibody product type   primary antibodies
UniProt accession no.   Q9Y6K8
Gene Information   human ... AK5(26289)

Description

General description

Adenylate kinase 5 (AK5), a member of adenylate kinase family is a nucleoside monophosphate kinase. It is localized on human chromosome 1p31.1. It is expressed in cytosol and abundant in the brain.

Immunogen

AK5 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Biochem/physiol Actions

Adenylate kinase 5 (AK5) plays a vital role in neuronal-specific metabolic processes and maintenance of cellular homeostasis. It also mediates the synthesis of DNA and RNA and energy transfer. AK5 is a phosphotransferase which catalyzes the interconversion of adenine nucleotides.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?