• USA Home
  • SAB4100342 - Monoclonal Anti-PSMB4 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4100342 Sigma-Aldrich

Monoclonal Anti-PSMB4 antibody produced in mouse

clone PSMB4-A291, culture supernatant

Synonym: Anti-HN3, Anti-HsN3, Anti-PROS-26, Anti-PROS26, Anti-Proteasome (prosome, macropain) subunit, β type, 4

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PS-PS, Primary Antibodies
conjugate   unconjugated
clone   PSMB4-A291, monoclonal
application(s)   western blot: suitable
species reactivity   human
mol wt   antigen predicted mol wt 29 kDa
form   aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   culture supernatant
isotype   IgG1
Quality Level   200
antibody product type   primary antibodies
UniProt accession no.   P28070
Gene Information   human ... PSMB4(5692)

Description

Immunogen

PSMB4 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?