• USA Home
  • SAB4200526 - Monoclonal Anti-FLI1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4200526 Sigma-Aldrich

Monoclonal Anti-FLI1 antibody produced in mouse

clone F4.23, tissue culture supernatant

Synonym: Anti-EWSR2, Anti-Friend leukemia integration 1 transcription factor, Anti-Friend leukemia virus integration 1, Anti-Proto-oncogene Fli-1, Anti-SIC-1, Anti-Transcription factor ERGB

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Gene Regulation and Expression, Antibodies to Transcription Regulators,
conjugate   unconjugated
clone   F4.23, monoclonal
biological source   mouse
application(s)   indirect immunofluorescence: 1:8,000-1-16,000 using HeLa cells.
  western blot: 1:16,000-1:32,000 using 3T3-L1 total cell extracts.
species reactivity   rat, monkey, bovine, mouse, dog, human
mol wt   antigen mol wt ~55 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   tissue culture supernatant
antibody product type   primary antibodies
UniProt accession no.   Q01543
Gene Information   human ... FLI1(2313)

Description

Immunogen

synthetic peptide corresponding to a sequence at the N-terminus of human FLI1, conjugated to KLH.

Physical form

Culture supernatant solution containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?