• USA Home
  • SAB4200850 - Anti-Proteus vulgaris LPS antibody, Mouse monoclonal

EMAIL THIS PAGE TO A FRIEND
SAB4200850 Sigma-Aldrich

Anti-Proteus vulgaris LPS antibody, Mouse monoclonal

clone P.vul-111, purified from hybridoma cell culture

Purchase

Properties

Related Categories Antibodies, Microbiome Antibodies More...
antibody form   purified from hybridoma cell culture
antibody product type   primary antibodies
clone   P.vul-111, monoclonal
description   Research area: Microbiome
species reactivity   Proteus vulgaris
packaging   antibody small pack of 25 μL
concentration   ~1 mg/mL
application(s)   immunoblotting: 0.05-01 μg/mL using using dead P. vulgaris bacteria lysate.
isotype   IgM
shipped in   dry ice
storage temp.   −20°C

Description

General description

Proteus vulgaris is a Gram negative rod-shaped bacteria, belongs to the Enterobacteriaceae family. Members of the genus Proteus (Proteus spp.) which also includes Proteus mirabilis, Proteus penneri and Proteus hauseri, originally characterize by their ability to swarm on solid surfaces, are widespread in the environment and the gastrointestinal tract of human and animals and known to be an opportunistic pathogens isolated from urine, wounds and other clinical sources. The Proteus spp. bacteria, are distinguished by their reactions for indole production, salicin fermentation and aesculin hydrolysis.1-3 P. vulgaris produces indole which differentiates it from the indole-negative P. mirabilis and P. penneri.1-3 Proteus spp. bacteria may also be found in soil or water habitats where they often regarded as indicators of fecal pollution and a contamination threat for potential water or seafood poisoning.1

Specificity

Monoclonal Anti-Proteus vulgaris LPS specifically recognizes P. vulgaris whole extract and P. vulgaris LPS, the antibody has no cross reactivity with whole extract of Proteus mirabilis, P. gingivalis, E. coli, Pseudomonas aeruginosa, Shigella flexneri, Staphylococcus aureus or Salmonella enterica.

Immunogen

Proteus vulgaris dead OX19 bacteria: ATCC strain 6380

Application

The antibody is recommended to be used in various immunological techniques, including Immunoblot and ELISA. Detection of the P. vulgaris band by Immunoblotting is specifically inhibited by the immunogen.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?