• USA Home
  • SAB4502649 - Anti-ROBO2 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4502649 Sigma-Aldrich

Anti-ROBO2 antibody produced in rabbit

affinity isolated antibody

Synonym: KIAA1568

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RO-RP
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   ELISA: 1:40000
  immunohistochemistry: 1:50-1:100
species reactivity   mouse, human, rat
mol wt   antigen 151 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   ~1 mg/mL
NCBI accession no.   Q9HCK4
UniProt accession no.   Q9HCK4
Gene Information   human ... ROBO2(6092)

Description

General description

Anti-ROBO2 Antibody detects endogenous levels of total ROBO2 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human ROBO2.

Immunogen Range: 237-286

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?