• USA Home
  • SAB4503273 - Anti-CASP1 (p20, Cleaved-Asn120) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4503273 Sigma-Aldrich

Anti-CASP1 (p20, Cleaved-Asn120) antibody produced in rabbit

affinity isolated antibody

Synonym: CASP-1, ICE, IL-1 β converting enzyme, IL-1BC, caspase-1

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CS-CS, Primary Antibodies
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   ELISA: 1:20000
  western blot: 1:500-1:1000
species reactivity   human
mol wt   antigen 18 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   ~1 mg/mL
NCBI accession no.   P29466
UniProt accession no.   P29466
Gene Information   human ... CASP1(834)

Description

General description

Anti-CSPN1 (p20, Cleaved-Asn120) Antibody detects endogenous levels of fragment of activated CSPN1 (p20, Cleaved-Asn120) protein.

Caspase-1 is also known as interleukin-1β converting enzyme (ICE). It is located on human chromosome 11q22.2. It has 10 exons, an N terminal CARD (caspase activation and recruitment domain), a large P20 subunit and a small P10 subunit. CASP1 is present in leukocytes, monocytes and epithelial cells.

Immunogen

The antiserum was produced against synthesized peptide derived from human Caspase 1.

Immunogen Range: 101-150

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Biochem/physiol Actions

Caspase-1 can transform pro-inflammatory cytokines, interleukin-1β (IL-1β) and IL-18 into their active forms. It participates in pyroptosis. It is overexpressed in the aorta of coronary atherosclerosis patients. CASP1 helps in host cell survival by inducing membrane biogenesis to restore the impairment caused by pore-forming toxins.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?