• USA Home
  • SAB4700602 - Anti-CD358-PE antibody, Mouse monoclonal

EMAIL THIS PAGE TO A FRIEND
SAB4700602 Sigma-Aldrich

Anti-CD358-PE antibody, Mouse monoclonal

clone DR-6-04-EC, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-DR6

  •  NACRES NA.41

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Flow Cytometry, Antibodies to CD and Related Antigens, Primary Antibodies,
conjugate   phycoerythrin (R-PE) conjugate
clone   DR-6-04-EC, monoclonal
biological source   mouse
application(s)   flow cytometry: suitable
species reactivity   human
form   buffered aqueous solution
shipped in   wet ice
storage temp.   2-8°C
antibody form   purified immunoglobulin
isotype   IgG1
antibody product type   primary antibodies
concentration   0.1 mg/mL
NCBI accession no.   NM_014452.3
UniProt accession no.   O75509
Gene Information   human ... TNFRSF21(27242)

Description

General description

The antibody DR-6-04-EC recognizes human DR6 (Death receptor-6), a type I transmembrane protein containing cytoplasmic death domain, widely expressed in most human tissues and cell lines.

Immunogen

A fusion protein representing amino acids 42-335 (extracellular part) of human DR6 linked to the Fc portion of human IgG1

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 10 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?