• USA Home
  • SAB4800014 - Imprint® Anti-HDAC1, (C-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4800014 Sigma-Aldrich

Imprint® Anti-HDAC1, (C-terminal) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-GON-10, Anti-HD1, Anti-HDAC1, Anti-RPD3, Anti-RPD3L1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Epigenetics and Nuclear Signaling, ChIP Validated Antibodies, H1-HE,
conjugate   unconjugated
clone   polyclonal
biological source   rabbit
application(s)   ChIP: suitable
  indirect ELISA: suitable
  western blot: suitable
species reactivity   human
mol wt   antigen ~55 kDa
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
antibody form   affinity isolated antibody
antibody product type   primary antibodies
concentration   ~0.64 mg/mL
NCBI accession no.   NM_004964
  NM_004964
UniProt accession no.   Q13547
Gene Information   human ... HDAC1(3065)

Description

Physical form

Supplied at 0.64 mg/mL in PBS, 0.05% sodium azide., 0.05% ProClin 300.

Legal Information

Imprint is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Histone Modification and Chromatin Remodeling

Gene expression is governed by complex mechanisms including transcription factor binding to DNA and coordinated changes in chromatin structure. The primary protein components of chromatin are the his...
Savita Bagga, PhD.
BioFiles v7 n3, 2012, 10–16
Keywords: Acetylations, Amplification, Cancer, Cell culture, Chromatin immunoprecipitation, Cloning, DNA purification, Diseases, Epigenetics, Gene expression, Immunoprecipitation, Indicators, Methylations, Microarray Analysis, Polymerase chain reaction, Polymerase chain reaction - quantitative, Polymorphisms, Purification, Sequencing, Transcription, Whole genome amplification

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detection methods, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?