• USA Home
  • SAB5202211 - Monoclonal Anti-Kv3.2-Alkaline Phosphatase antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB5202211 Sigma-Aldrich

Monoclonal Anti-Kv3.2-Alkaline Phosphatase antibody produced in mouse

clone S410-17

  •  NACRES NA.45

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, KL-KY, Primary Antibodies
conjugate   alkaline phosphatase conjugate
clone   S410-17, monoclonal
biological source   mouse
application(s)   immunofluorescence: 1:100
  western blot: 1:1000
species reactivity   mouse, human
mol wt   90 kDa
form   buffered aqueous solution
shipped in   wet ice
storage temp.   −20°C
antibody form   purified immunoglobulin
isotype   IgG1
antibody product type   primary antibodies
concentration   1.0 mg/mL
NCBI accession no.   NP_631962.1
Gene Information   rat ... Kcnc2(246153)

Description

Immunogen

Fusion protein amino acids 474-613 (Cytoplasmic C-terminus) of rat Kv3.2a

Physical form

PBS pH 7.4, 50% glyercol, 0.09% Sodium Azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?