• USA Home
  • SAB5300318 - Monoclonal Anti-CSF1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB5300318 Sigma-Aldrich

Monoclonal Anti-CSF1 antibody produced in mouse

clone 2D10, ascites fluid

Synonym: MCSF

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, CS-CS, Primary Antibodies
conjugate   unconjugated
clone   2D10, monoclonal
biological source   mouse
application(s)   direct ELISA: 1:10,000
  immunohistochemistry: 1:200-1:1,000
  western blot: 1:500-1:2,000
species reactivity   human
mol wt   60 kDa
shipped in   wet ice
storage temp.   −20°C
antibody form   ascites fluid
isotype   IgG1
antibody product type   primary antibodies
NCBI accession no.   NM_000757
UniProt accession no.   P09603
Gene Information   human ... CSF1(1435)

Description

Immunogen

Purified recombinant fragment of human CSF1 expressed in E.coli.
Mouse monoclonal antibody raised against CSF1

Physical form

Ascitic fluid containing 0.03% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Protocols

Western Blot Protocol | Immunoblotting Protocol

Western Blotting refers to the electrophoretic transfer of proteins from sodium dodecyl sulfate polyacrylamide gels to sheets of PVDF or nitrocellullose membrane, followed by immunodetection of prote...
Keywords: AGE, Buffers, Cell disruption, Detergents, Dialysis, Electroblotting, Electrophoresis, Enzyme activity, Gel electrophoresis, Immunoprecipitation, PAGE, Protein extraction, Purification, Sample preparations, Western blot

Related Content

Antibody Explorer | Buy Primary Antibodies & Secondary Antibodies

Primary, Secondary and Recombinant Monoclonal Antibodies Providing highly cited primary and secondary antibodies, we have you covered for your ELISA, western blot, immunohistochemistry or other assay...
Keywords: Alzheimer Disease, Cancer, Chromatin immunoprecipitation, Diagnostic, Enzyme-linked immunosorbent assay, Flow cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Neuroscience, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?