• USA Home
  • SBP2505 - Livin Blocking Peptide for PRS2505

EMAIL THIS PAGE TO A FRIEND
SBP2505 Sigma-Aldrich

Livin Blocking Peptide for PRS2505

Purchase

Properties

Quality Level   100
biological source   synthetic
form   liquid
species reactivity   human
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Immunogen

35 amino acids of human Livin.

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody PRS2505.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?