• USA Home
  • SBP3500021 - PCDH18 Blocking Peptide for SAB3500021

EMAIL THIS PAGE TO A FRIEND
SBP3500021 Sigma-Aldrich

PCDH18 Blocking Peptide for SAB3500021

Synonym: PCDH18 Blocking Peptide for SAB3500021

  •  NACRES NA.41

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   rat, mouse, human
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Immunogen

18 amino acids near the amino terminus of human PCDH18.

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody SAB3500021.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SAB3500021 Anti-PCDH18 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?