• USA Home
  • SBP3501013 - TOX Blocking Peptide for SAB3501013

EMAIL THIS PAGE TO A FRIEND
SBP3501013 Sigma-Aldrich

TOX Blocking Peptide for SAB3501013

Synonym: TOX Blocking Peptide for SAB3501013

  •  NACRES NA.41

Purchase

Properties

biological source   synthetic
form   liquid
species reactivity   human
concentration   200 μg/mL
shipped in   wet ice
storage temp.   −20°C

Description

Immunogen

17 amino acid peptide near the amino terminus of human TOX.

Application

Used as a blocking peptide in immunoblotting applications.

Linkage

This peptide blocks the action of antibody SAB3501013.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

SAB3501013 Anti-TOX antibody produced in rabbit, affinity isolated antibody

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?