• USA Home
  • SBPV05 - MRP5 Human Vesicular Transport Kit

EMAIL THIS PAGE TO A FRIEND
SBPV05 Sigma-Aldrich

MRP5 Human Vesicular Transport Kit

membrane based kit for Human MRP5 Vesicular Transport Assays

Synonym: ABCC5, Multidrug resistance associated protein 5

Purchase

Properties

recombinant   expressed in human cells
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... ABCC5(10057)

Description

General description

SOLVO Biotechnology offers ready-to-use assay kits for efflux and uptake transporters. SOLVO′s PREDIVEZ Vesicular Transport Assay Kits are suitable for efflux transporter inhibition studies. The kits are designed to test the inhibitory effect of one or more test

Application

The PREDIVEZTM Vesicular Transport Kit is a simple and powerful tool to investigate drug transporter interactions. It provides information on any interaction between the ABC transporter and the drug candidate that would affect the transport of the fluorescent or radiolabeled or cold.

PREDIVEZTM vesicular transport kits are available in three sizes 3, 6 or 9 compound sizes. The kits contain the sufficient amount of reagents and membrane preparations needed for the testing of 3 compounds per 96-well plate. Each kit is accompanied by a CD that includes the assay protocol, an Excel spreadsheet with a data processing template, SDS and the inhibition curves with reference compounds.

Physical form

Supplied as frozen membrane vesicles, containing 5 mg/ml membrane protein, labeled with volume, catalog number (transporter), date of production, protocols and necessary reagents sufficent for the analysis of 3, 6, or 9 test compounds

Legal Information

Distributed for SOLVO Biotechnology, Inc.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1824 8 / PGIII

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

SBCT06 SB-HEK293-Mock-CTRL, Negative control membrane for HEK293 transporter membranes
SBVT08 SB-MRP5-HEK293, MRP5 human membrane vesicles

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?