EMAIL THIS PAGE TO A FRIEND
SCP0128 Sigma-Aldrich

DRONC Substrate I

  • Empirical Formula (Hill Notation) C30H37N6O12

  • Molecular Weight 673.65

  •  NACRES NA.32

Purchase

Properties

Related Categories Caspase Inhibitors and Substrates, Caspase Substrates, Cell Biology, Cell Signaling and Neuroscience, DRONC,
assay   ≥95% (HPLC)
form   lyophilized
composition   Peptide Content, ≥85%
storage condition   protect from light
storage temp.   −20°C

Description

Amino Acid Sequence

Ac-Thr-Gln-Thr-Glu-AFC

Application

Ac-TQTE-AFC (Ac-Thr-Gln-Thr-Glu-AFC) and Ac-TQTD-AFC (Ac-Thr-Gln-Thr-Asp-AFC) are fluorogenic substrates use to detect and identify the DRONC caspase(s).

Safety & Documentation

Safety Information

WGK Germany 
-

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?