EMAIL THIS PAGE TO A FRIEND
SIHK0539 Sigma-Aldrich

MISSION® siRNA

Human Kinase DAPK1 (siRNA2), Nano Scale 0.25 nmol

Purchase

Properties

product line   MISSION®
storage condition   desiccated
NCBI accession no.   NM_004938
storage temp.   −20°C
Gene Information   human ... DAPK1(1612)

Description

General description

The MISSION small scale siRNA are available at 0.25 nmol scale. Sigma′s siRNAs are designed with the Rosetta algorithm and are backed by Sigma-Aldrich’s predesigned siRNA guarantee. You can find all of these off-the-shelf siRNAs on YFG.

Other Notes

Associated siRNA: SASI_Hs01_00238226

Physical form

lyophilized powder

Legal Information

MISSION is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?