EMAIL THIS PAGE TO A FRIEND
SML0145 Sigma-Aldrich

MRT-10

≥98% (HPLC)

Synonym: N-[[[3-[(Benzoyl)amino]phenyl]amino](thioxo)methyl]-3,4,5-trimethoxybenzamide

Purchase

Properties

Related Categories Angiogenic Factors, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
InChI Key   KVQVEZQDNHMQJV-UHFFFAOYSA-N
assay   ≥98% (HPLC)
form   powder
storage condition   desiccated
color   white to tan
solubility   DMSO: ≥15 mg/mL
storage temp.   2-8°C

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

MRT-10 is a novel, high-affinity antagonist of smoothened (Smo). MRT-10 displaces labeled cyclopamine from Smo with an IC50 of 500 nM, and inhibits shh-induced Gli-reporter activity in Shh-light2 cells (IC50 = 0.64 μM).

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number

Milli-Q® Water Purification Solutions
Protocols & Articles

Articles

Discover Bioactive Small Molecules for Cytokines & Growth Factors Research

Cytokines and growth factors (GF) are chemical messengers that mediate intracellular communication, thereby regulating cellular and nuclear functions. These chemical messengers bind cell surface rece...
Keywords: Biological processes, Cancer, Cell proliferation, Diseases, Growth factors, Ligands, Phosphorylations, Transcription, Transduction

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?