EMAIL THIS PAGE TO A FRIEND
SML0867 Sigma-Aldrich

BG45

≥98% (HPLC)

Synonym: N-(2-Aminophenyl)pyrazine-2-carboxamide

  • CAS Number 926259-99-6

  • Empirical Formula (Hill Notation) C11H10N4O

  • Molecular Weight 214.22

  •  NACRES NA.77

Purchase

Properties

Related Categories B, Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience,
Quality Level   100
assay   ≥98% (HPLC)
form   powder
color   white to beige
solubility   DMSO: 20 mg/mL, clear
storage temp.   2-8°C

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

BG45 is an HDAC class I inhibitor with selectivity for HDAC3 (IC50 = 289 nM) over HDAC1, 2. BG45 did not inihibit HDAC6. BG45 signigicantly inhibited tumor growth in a mouse model of multiple myeloma either alone and synergistically in combination with bortezomib.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Discover Bioactive Small Molecules for Gene Regulation

The loss of regulation of gene expression is a key component to many human disease states, including neurodegenerative disorders, autoimmune conditions and, most prominently, cancers. Regulation of g...
Keywords: Acetylations, Epigenetics, Gene expression, Genomics, Methylations, PAGE, Post translational modifications, Transcription

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?