EMAIL THIS PAGE TO A FRIEND
SML1627 Sigma-Aldrich

DFHBI

Synonym: (5Z)-5-[(3,5-Difluoro-4-hydroxyphenyl)methylene]-3,5-dihydro-2,3-dimethyl- 4H-Imidazol-4-one, (Z)-4-(3,5-Difluoro-4-hydroxybenzylidene)-1,2-dimethyl-1H-imidazol-5(4H)-one

  • CAS Number 1241390-29-3

  • Empirical Formula (Hill Notation) C12H10F2N2O2

  • Molecular Weight 252.22

  •  PubChem Substance ID 329825511

  •  NACRES NA.77

Purchase

Properties

Related Categories Bioactive Small Molecule Alphabetical Index, Bioactive Small Molecules, Cell Biology, Cell Signaling and Neuroscience, D-DIF,
Quality Level   100
assay   ≥98% (HPLC)
form   powder
color   yellow to orange
solubility   DMSO: 20 mg/mL, clear
storage temp.   −20°C
SMILES string   FC1=CC(/C=C2N=C(C)N(C)C\2=O)=CC(F)=C1O
InChI   1S/C12H10F2N2O2/c1-6-15-10(12(18)16(6)2)5-7-3-8(13)11(17)9(14)4-7/h3-5,17H,1-2H3/b10-5-
InChI key   ZDDIJYXDUBFLID-YHYXMXQVSA-N

Description

Packaging

5, 25 mg in glass bottle

Biochem/physiol Actions

3,5-difluoro-4-hydroxybenzylidene imidazolinone (DFHBI) is non fluorescent when unbound.

DFHBI is a membrane permeable and nontoxic fluorogen, which produces fluorescence upon binding. RNA aptamers Spinach, Spinach2 and Broccoli. DFHBI displays fluorescence excitation maxima of 447 nm and peak fluorescence emission of 501 nm. DFHBI is used for imaging RNA in cells.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Target organs 
Respiratory system
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?