• USA Home
  • SRE0032 - Tris Buffered Saline Solution

EMAIL THIS PAGE TO A FRIEND
SRE0032 Sigma-Aldrich

Tris Buffered Saline Solution

For in-vitro Diagnostic Use

Synonym: TBS, Tris Buffered Saline

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions
Quality Level   GMP-IVD
form   liquid
application(s)   ELISA: suitable
  immunohistochemistry: suitable
  western blot: suitable
Featured Industry   Diagnostic Assay Manufacturing
shipped in   wet ice
storage temp.   2-8°C

Description

Application

For use as a wash buffer in western blotting, ELISA, and immunohistochemical (IHC) staining procedures.

Suitability

Suitable for use in diagnostic applications and general use lab applications such as Dot Blot, western blotting, enzyme-linked immunosorbent assay (ELISA), and immunohistochemical (IHC) staining procedures.

Physical form

10X Solution as provided: 0.5 M Tris/Tris-HCl, 1.38 M NaCl, and 0.027 M KCl in high purity dH2O adjusted to pH 7.4. Solution is 0.2 μm filtered.

1X Working solution will consist of 0.05 M Tris/Tris-HCl, 0.138 M NaCl, and 0.0027 M KCl, pH 7.3 - 7.5, at 25 °C.

Other Notes

Concentrated solution should be stored at 2–8 °C. Precaution should be taken to minimize foaming. Avoid vigorous agitation. The diluted wash buffer can be stored at room temperature (20–25 °C) for up to one week. Solutions that become turbid should be discarded.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Buffer Solutions

Product development in IVD relies on the quality and performance of raw materials that constitute the basic components of any reagent/kit. Buffers constitute a major critical reagent and help stabili...
Keywords: Buffers, Diagnostic

ISO 13485 vs. CFR820 White Paper

For vendor qualification requirements see: PROC-GLOB-POL-024275 For appropriate qualifier/extended name suffix statements see: PROC-GLOB-REF-023112 For appropriate label disclaimer/usage statements s...
Keywords: Capillary electrophoresis

ISO 13485 vs. ISO 9001

Misunderstandings about International Organization for Standardization (ISO) management standard 13485, and how it compares to other industry standards such as ISO 9001 or Current Good Manufacturing ...
Keywords: Addition reactions, Clinical, Diagnostic, Hematology, Polymerase chain reaction - quantitative, Purification

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?