• USA Home
  • SRE0034 - Carbonate Bicarbonate Buffer

EMAIL THIS PAGE TO A FRIEND
SRE0034 Sigma-Aldrich

Carbonate Bicarbonate Buffer

For in-vitro Diagnostic Use

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions
Quality Level   GMP-IVD
form   powder
Featured Industry   Diagnostic Assay Manufacturing
shipped in   wet ice
storage temp.   2-8°C

Description

Application

Na-carbonate (carb-bicarb) buffer for use in protein and antibody coating procedures.

Suitability

Suitable for use as a coating buffer for protein-coating procedures on microtiter plates and plastic tubes for radioimmunoassay (RIA) and enzyme immunoassay (EIA) techniques

Physical form

Product is a preformulated powder to be dissolved for use by diagnostic or clinical laboratories or by manufacturers of diagnostic reagents or kits. Prepare the Single Use Disposable Solution (SUDS) by dissolving the powder in 1L of purified water for a 1X working solution of 0.05 M sodium carbonate-bicarbonate buffer with pH 9.6 at 25 °C. Approximately 70:30 bicarb:carb ratio.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Certificate of Origin

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Buffer Solutions

Product development in IVD relies on the quality and performance of raw materials that constitute the basic components of any reagent/kit. Buffers constitute a major critical reagent and help stabili...
Keywords: Buffers, Diagnostic

ISO 13485 vs. CFR820 White Paper

For vendor qualification requirements see: PROC-GLOB-POL-024275 For appropriate qualifier/extended name suffix statements see: PROC-GLOB-REF-023112 For appropriate label disclaimer/usage statements s...
Keywords: Capillary electrophoresis

ISO 13485 vs. ISO 9001

Misunderstandings about International Organization for Standardization (ISO) management standard 13485, and how it compares to other industry standards such as ISO 9001 or Current Good Manufacturing ...
Keywords: Addition reactions, Clinical, Diagnostic, Hematology, Polymerase chain reaction - quantitative, Purification

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?