• USA Home
  • SRE0063 - Tris-EDTA buffer solution, pH 9.0

EMAIL THIS PAGE TO A FRIEND
SRE0063 Sigma-Aldrich

Tris-EDTA buffer solution, pH 9.0

For in-vitro Diagnostic Use

Synonym: Tris-EDTA buffer

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions
Quality Level   GMP-IVD
form   liquid
Featured Industry   Diagnostic Assay Manufacturing
shipped in   wet ice
storage temp.   2-8°C

Description

Application

For use in Diagnostic applications for the heat induced epitope (or antigen) retrieval (HIER) of formalin-fixed, paraffin-embedded (FFPE) tissues prior to immunohistochemical (IHC) staining.

Preformulated buffer for use by diagnostic or clinical laboratories or by manufacturers of diagnostic reagents or kits.

Suitability

Preformulated buffer for use by diagnostic or clinical laboratories or by manufacturers of diagnostic reagents or kits.

Physical form

This Ready-to-use Tris-EDTA buffer contains 10mM Trizma® Base with 1mm EDTA at pH 9.

Other Notes

Precautions: Excessive epitope retrieval of FFPE tissues could result in damage of tissue morphology or tissue sections becoming detached from the slide. Inadequate epitope retrieval of FFPE tissue could result in weaker staining. Normal precautions exercised in handling laboratory reagents should be followed

Legal Information

Trizma is a registered trademark of Sigma-Aldrich Co. LLC

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?