• USA Home
  • SRE0064 - Phosphate Buffer Solution, 1M

EMAIL THIS PAGE TO A FRIEND
SRE0064 Sigma-Aldrich

Phosphate Buffer Solution, 1M

For in-vitro Diagnostic Use

Synonym: Phosphate Buffer Solution

Purchase

Properties

Related Categories Biochemicals and Reagents, Biological Buffers, Buffer Convenience Packaging, Buffer Solutions
Quality Level   GMP-IVD
form   liquid
Featured Industry   Diagnostic Assay Manufacturing

Description

Application

Intended for use as a buffer in various Romanowsky type staining procedures, including Wright Stain, Wright Giemsa, Giemsa, May Grunwald, Jenner and Leishman. For use in the staining and visualization of structures and/or other intra/extracellular elements in biological tissues.

Suitability

Preformulated buffer for use by diagnostic or clinical laboratories or by manufacturers of diagnostic reagents or kits.

Physical form

This product is a ready-to-use 1M Phosphate Buffer solution containing sodium phosphate dibasic and potassium phosphate monobasic. The pH of the product is adjusted using hydrochloric acid and sodium hydroxide.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Related Content

BioUltra Biological Buffers

A buffer, as defined by Van Slyke [1], is "a substance which by its presence in solution increases the amount of acid or alkali that must be added to cause unit change in pH". Buffers are thus very i...
Keywords: Absorption, Adsorption, Biochemistry, Biological Buffers, Biological processes, Buffers, PAGE

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?