EMAIL THIS PAGE TO A FRIEND
SRP0358 Sigma-Aldrich

Akt3 active human

recombinant, expressed in baculovirus infected Sf9 cells, ≥90% (SDS-PAGE)

Synonym: PKBG, PRKBG, v-akt murine thymoma viral oncogene homolog 3 (protein kinase B, gamma)

  •  NACRES NA.32

Purchase

Properties

Related Categories Application Index, Biochemicals and Reagents, Cell Biology, Cell Signaling Enzymes, Cell Signaling and Neuroscience,
biological source   human
recombinant   expressed in baculovirus infected Sf9 cells
assay   ≥90% (SDS-PAGE)
form   aqueous solution
mol wt   60 kDa
packaging   pkg of 10 μg
storage condition   avoid repeated freeze/thaw cycles
NCBI accession no.   NM_005465
UniProt accession no.   Q9Y243
shipped in   dry ice
storage temp.   −70°C
Gene Information   human ... AKT3(10000)

Description

General description

Recombinant human AKT3 (GenBank Accession No. NM_005465), amino acids 2-end with N-terminal HIS-tag, MW= 60 kDa, expressed in Baculovirus expression system.

Application

Useful for the study of enzyme kinetics, screening inhibitors, and selectivity profiling.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 1
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?