• USA Home
  • SRP3220 - Lungkine from mouse

EMAIL THIS PAGE TO A FRIEND
SRP3220 Sigma-Aldrich

Lungkine from mouse

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), cell culture tested

Synonym: CXCL15

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Chemokines, Cytokines and Growth Factors, Cytokines, Growth Factors and Chemokines for Stem Cell Differentiation,
biological source   mouse
recombinant   expressed in E. coli
assay   ≥98% (HPLC)
  ≥98% (SDS-PAGE)
form   lyophilized
mol wt   16.3 kDa
packaging   pkg of 20 μg
impurities   <0.1 EU/μg endotoxin, tested
color   white to off-white
suitability   cell culture tested
UniProt accession no.   Q9WVL7
shipped in   wet ice
storage temp.   −20°C
Gene Information   mouse ... CXCL15(20309)

Description

General description

Lungkine is a CXC chemokine that is expressed in lung epithelial cells and, to a lesser extent, in certain fetal tissues. No human homolog has been identified and a specific cell surface receptor has not yet been found. Lungkine expression in lung tissue is elevated in response to inflammation, at which time it acts to specifically recruit neutrophils and direct them into the lung airway. Recombinant murine Lungkine is a 16.3 kDa CXC chemokine consisting of 142 amino acid residues, including four conserved cysteine residues present in CXC chemokines.

Biochem/physiol Actions

Lungkine is a CXC chemokine that is expressed in lung epithelial cells and, to a lesser extent, in certain fetal tissues. Recombinant murine Lungkine is a 16.3 kDa CXC chemokine consisting of 142 amino acid residues, including four conserved cysteine residues present in CXC chemokines.

Sequence

QELRCLCIQE HSEFIPLKLI KNIMVIFETI YCNRKEVIAV PKNGSMICLD PDAPWVKATV GPITNRFLPE DLKQKEFPPA MKLLYSVEHE KPLYLSFGRP ENKRIFPFPI RETSRHFADL AHNSDRNFLR DSSEVSLTGS DA

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Request a Serum Sample

Milli-Q® Water Purification Solutions
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?