EMAIL THIS PAGE TO A FRIEND
SRP4063 Sigma-Aldrich

FGF-22 human

recombinant, expressed in E. coli, ≥97% (SDS-PAGE), ≥97% (HPLC)

Synonym: FGF22, Fibroblast growth factor 22

Purchase

Properties

Related Categories Cell Biology, Cell Culture, Cell Signaling and Neuroscience, Cytokines, Cytokines and Growth Factors,
biological source   human
recombinant   expressed in E. coli
assay   ≥97% (HPLC)
  ≥97% (SDS-PAGE)
form   lyophilized
mol wt   mol wt ~17.3 kDa
packaging   pkg of 10 μg
storage condition   avoid repeated freeze/thaw cycles
impurities   endotoxin, tested
NCBI accession no.   NM_020637
UniProt accession no.   Q9HCT0
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... FGF22(27006)

Description

General description

Fibroblast Growth Factor-22 stimulates the proliferation and activation of cells that express FGF receptor. The human FGF-22 is a 17.3 kDa protein containing 149 amino acid residues.

Reconstitution

Centrifuge the vial prior to opening. Avoid freeze-thaw cycles.

Reconstitute in sterile water to a concentration no less than 100 μg/ml of FGF-22.

Physical form

Sterile filtered and Lyophilized without additives.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis


Request a Serum Sample

Cell Culture Guide
Protocols & Articles

Articles

Fibroblast Growth Factor Family (FGF)

Fibroblast Growth Factors (FGFs) are potent regulators of cell proliferation, differentiation and function and are critically important in normal development, tissue maintenance, wound repair and ang...
Keywords: Angiogenesis, Cancer, Cardiovascular, Cell proliferation, Diabetes, Diseases, Gene expression, Growth factors, Ligands

Related Content

Cell Culture Troubleshooting Guide

Cell culture has become one of the most fundamental techniques for modeling biological systems, and is of increasing importance in the biotechnology and pharmaceutical sectors as well as an essential...
Keywords: Antibiotics, Apoptosis, Cell attachment, Cell culture, Cryopreservation, Filtration, Growth factors, Pharmaceutical, Phase transitions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?